Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093669.4 | XP_001093669.1 | 3199 | 60 | ||
XM_028828514.1 | XP_028684347.1 | 1884 | 60 | |||
GenBank mRNA | JV066365.1 | - | 1241 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |