Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266838.1 | NP_001253767.1 | 811 | 71 | ||
XM_015134895.2 | XP_014990381.1 | 1062 | 71 | |||
XM_015134896.2 | XP_014990382.1 | 818 | 71 | |||
XM_015134898.2 | XP_014990384.1 | 3024 | 71 | |||
XM_015134899.2 | XP_014990385.1 | 831 | 71 | |||
XM_015134901.2 | XP_014990387.1 | 682 | 71 | |||
XM_028844884.1 | XP_028700717.1 | 680 | 71 | |||
GenBank mRNA | JU321044.1 | - | 811 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |