Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257592.1 | NP_001244521.1 | 1031 | 94 | ||
XM_028843267.1 | XP_028699100.1 | 1312 | 94 | |||
GenBank mRNA | JU320493.1 | - | 1031 | 94 | ||
JV043989.1 | - | 1070 | 94 | |||
JV529168.1 | - | 1103 | 94 | |||
JV566819.1 | - | 1091 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |