Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261709.1 | NP_001248638.1 | 1103 | 94 | ||
XM_015128604.2 | XP_014984090.1 | 1167 | 94 | |||
XM_015128608.2 | XP_014984094.1 | 1646 | 94 | |||
XM_015128611.2 | XP_014984097.1 | 1385 | 94 | |||
XM_015128617.2 | XP_014984103.1 | 1164 | 94 | |||
XM_015128629.2 | XP_014984115.1 | 1643 | 94 | |||
GenBank mRNA | JU336781.1 | - | 1132 | 94 | ||
JV048481.1 | - | 1103 | 94 |