Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260620.1 | NP_001247549.1 | 575 | 79 | ||
GenBank mRNA | JU471273.1 | - | 575 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |