Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257745.1 | NP_001244674.1 | 1312 | 53 | ||
GenBank mRNA | JU320309.1 | - | 1312 | 53 | ||
JU473342.1 | - | 1175 | 53 | |||
JV044752.1 | - | 1042 | 53 | |||
JV667667.1 | - | 1325 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |