Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMMECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015128010.2 | XP_014983496.1 | 948 | 96 | ||
GenBank mRNA | JU497254.1 | - | 501 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |