Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261686.1 | NP_001248615.1 | 467 | 102 | ||
XM_015149019.2 | XP_015004505.1 | 763 | 102 | |||
XM_028827224.1 | XP_028683057.1 | 763 | 102 | |||
GenBank mRNA | JU335727.1 | - | 451 | 102 | ||
JU473122.1 | - | 467 | 102 | |||
JV637277.1 | - | 660 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |