Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266912.1 | NP_001253841.1 | 595 | 101 | ||
XM_015151807.2 | XP_015007293.1 | 589 | 101 | |||
XM_015151808.2 | XP_015007294.1 | 740 | 101 | |||
GenBank mRNA | JU319960.1 | - | 588 | 101 | ||
JU319961.1 | - | 588 | 101 | |||
JU332138.1 | - | 534 | 101 | |||
JU332139.1 | - | 588 | 101 | |||
JU474940.1 | - | 565 | 101 | |||
JV043983.1 | - | 595 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |