Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118714.2 | XP_014974200.2 | 2056 | 73 | ||
XM_015118715.2 | XP_014974201.2 | 2060 | 73 | |||
XM_028836054.1 | XP_028691887.1 | 2065 | 73 | |||
GenBank mRNA | JU325778.1 | - | 420 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |