Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033022.1 | NP_001028194.1 | 1664 | 55 | ||
NM_001033023.1 | NP_001028195.1 | 1799 | 55 | |||
GenBank mRNA | AF286026.1 | - | 1799 | 55 | ||
AF286027.1 | - | 1664 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |