Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261815.2 | NP_001248744.2 | 1121 | 65 | ||
XM_028844747.1 | XP_028700580.1 | 1753 | 65 | |||
XM_028844748.1 | XP_028700581.1 | 1754 | 65 | |||
XM_028844749.1 | XP_028700582.1 | 1748 | 65 | |||
XM_028844750.1 | XP_028700583.1 | 1754 | 65 | |||
XM_028844751.1 | XP_028700584.1 | 1684 | 65 | |||
XM_028844752.1 | XP_028700585.1 | 1360 | 65 | |||
XM_028844753.1 | XP_028700586.1 | 1377 | 65 | |||
XM_028844754.1 | XP_028700587.1 | 1752 | 65 | |||
XM_028844755.1 | XP_028700588.1 | 1360 | 65 | |||
XM_028844756.1 | XP_028700589.1 | 1535 | 65 | |||
XM_028844757.1 | XP_028700590.1 | 1145 | 65 | |||
XM_028844758.1 | XP_028700591.1 | 1360 | 65 | |||
XM_028844759.1 | XP_028700592.1 | 1360 | 65 | |||
GenBank mRNA | AY950560.1 | - | 85 | 65 | ||
JU327588.1 | - | 1092 | 65 | |||
JU327589.1 | - | 1086 | 65 | |||
JU471703.1 | - | 1125 | 65 | |||
JU474451.1 | - | 1125 | 65 | |||
JV566287.1 | - | 1166 | 65 | |||
JV635975.1 | - | 1185 | 65 | |||
JV635976.1 | - | 1179 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |