Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042433.2 | NP_001035898.1 | 1740 | 89 | ||
GenBank mRNA | DQ644021.1 | - | 1743 | 89 | ||
EF467994.1 | - | 1778 | 89 | |||
JU470652.1 | - | 1947 | 89 |