Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBP-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001096399.4 | XP_001096399.1 | 415 | 138 | ||
GenBank mRNA | L43912.1 | - | 290 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |