Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCPIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260931.1 | NP_001247860.1 | 2926 | 104 | ||
GenBank mRNA | JU327623.1 | - | 2563 | 104 | ||
JU477286.1 | - | 2926 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |