Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257836.1 | NP_001244765.1 | 359 | 110 | ||
XM_028835501.1 | XP_028691334.1 | 437 | 110 | |||
XM_028835502.1 | XP_028691335.1 | 437 | 110 | |||
XM_028835506.1 | XP_028691339.1 | 450 | 110 | |||
GenBank mRNA | JU333254.1 | - | 359 | 110 | ||
JU471579.1 | - | 365 | 110 | |||
JV046352.1 | - | 347 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |