Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266939.1 | NP_001253868.1 | 45 | 58 | ||
XM_015112736.2 | XP_014968222.2 | 109 | 58 | |||
XM_015112737.2 | XP_014968223.1 | 143 | 58 | |||
XM_015112738.2 | XP_014968224.1 | 112 | 58 | |||
XM_015112739.2 | XP_014968225.1 | 97 | 58 | |||
XM_015112740.2 | XP_014968226.1 | 54 | 58 | |||
GenBank mRNA | JU321163.1 | - | 40 | 58 | ||
JU472483.1 | - | 45 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |