Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266363.1 | NP_001253292.1 | 376 | 103 | ||
XM_015143706.2 | XP_014999192.1 | 508 | 103 | |||
GenBank mRNA | JU334469.1 | - | 423 | 103 | ||
JU475566.1 | - | 376 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |