Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194343.2 | NP_001181272.1 | 752 | 68 | ||
GenBank mRNA | JU323207.1 | - | 798 | 68 | ||
JU471549.1 | - | 733 | 68 | |||
JV045400.1 | - | 722 | 68 | |||
JV600534.1 | - | 826 | 68 | |||
JV637676.1 | - | 770 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |