Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083465.4 | XP_001083465.1 | 150 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |