Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194529.2 | NP_001181458.2 | 639 | 144 | ||
GenBank mRNA | JU325587.1 | - | 163 | 144 | ||
JV661683.1 | - | 533 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |