Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEX3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001110186.4 | XP_001110186.1 | 680 | 97 | ||
GenBank mRNA | JV663261.1 | - | 212 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |