Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEXIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119778.1 | XP_014975264.1 | 597 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |