Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002806358.3 | XP_002806404.1 | 1356 | 86 | ||
XM_015128025.2 | XP_014983511.1 | 1341 | 86 | |||
XM_015128026.2 | XP_014983512.1 | 1316 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |