Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP27B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194713.2 | NP_001181642.2 | 1135 | 89 | ||
XM_015152067.2 | XP_015007553.2 | 1534 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |