Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFTPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257727.1 | NP_001244656.1 | 204 | 126 | ||
XM_015112495.2 | XP_014967981.1 | 269 | 126 | |||
GenBank mRNA | JU332285.1 | - | 204 | 126 | ||
JU332286.1 | - | 204 | 126 | |||
JU472339.1 | - | 259 | 126 | |||
JU472340.1 | - | 259 | 126 | |||
JV044200.1 | - | 185 | 126 | |||
JV566312.1 | - | 268 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |