Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127873.2 | XP_014983359.1 | 160 | 55 | ||
XM_028841674.1 | XP_028697507.1 | 160 | 55 | |||
GenBank mRNA | JU477871.1 | - | 57 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |