Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130128.2 | XP_014985614.1 | 334 | 80 | ||
XM_015130129.2 | XP_014985615.1 | 340 | 80 | |||
XM_015130131.2 | XP_014985617.1 | 334 | 80 | |||
XM_028843187.1 | XP_028699020.1 | 486 | 80 | |||
GenBank mRNA | JU320319.1 | - | 239 | 80 | ||
JU320320.1 | - | 239 | 80 | |||
JU473049.1 | - | 139 | 80 | |||
JU473050.1 | - | 139 | 80 | |||
JV645848.1 | - | 242 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |