Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148162.2 | XP_015003648.2 | 1177 | 87 | ||
GenBank mRNA | JU332724.1 | - | 756 | 87 | ||
JU479402.1 | - | 659 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |