Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113386.4 | XP_001113386.1 | 362 | 82 | ||
GenBank mRNA | JU323417.1 | - | 282 | 82 | ||
JU473382.1 | - | 294 | 82 | |||
JV045576.1 | - | 298 | 82 | |||
JV529782.1 | - | 251 | 82 |