Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REC114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093604.4 | XP_001093604.1 | 189 | 152 | ||
XM_015142681.2 | XP_014998167.1 | 188 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |