Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266748.1 | NP_001253677.1 | 1282 | 127 | ||
XM_015131264.2 | XP_014986750.1 | 1687 | 127 | |||
XM_015131265.2 | XP_014986751.1 | 1641 | 127 | |||
XM_015131266.2 | XP_014986752.1 | 1331 | 127 | |||
GenBank mRNA | JU322566.1 | - | 1282 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |