Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257605.2 | NP_001244534.1 | 519 | 96 | ||
XM_028853315.1 | XP_028709148.1 | 620 | 96 | |||
XM_028853316.1 | XP_028709149.1 | 575 | 96 | |||
GenBank mRNA | JU323468.1 | - | 519 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |