Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266114.1 | NP_001253043.1 | 359 | 123 | ||
XM_015141273.2 | XP_014996759.2 | 503 | 123 | |||
XM_028849079.1 | XP_028704912.1 | 222 | 123 | |||
GenBank mRNA | JU336194.1 | - | 359 | 123 | ||
JV695120.1 | - | 385 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |