Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261059.2 | NP_001247988.1 | 271 | 137 | ||
GenBank mRNA | JU319855.1 | - | 175 | 137 | ||
JV048144.1 | - | 272 | 137 | |||
JV660459.1 | - | 548 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |