Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261693.1 | NP_001248622.1 | 677 | 90 | ||
GenBank mRNA | JU336890.1 | - | 672 | 90 | ||
JU477631.1 | - | 677 | 90 | |||
JV664368.1 | - | 433 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |