Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260905.1 | NP_001247834.1 | 1328 | 65 | ||
XM_015150010.2 | XP_015005496.1 | 1467 | 65 | |||
XM_015150011.2 | XP_015005497.1 | 1379 | 65 | |||
XM_028827420.1 | XP_028683253.1 | 1134 | 65 | |||
XM_028827421.1 | XP_028683254.1 | 1004 | 65 | |||
XM_028827422.1 | XP_028683255.1 | 991 | 65 | |||
GenBank mRNA | JU337091.1 | - | 1328 | 65 | ||
JV701220.1 | - | 1399 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |