Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261336.1 | NP_001248265.1 | 681 | 103 | ||
XM_015131358.2 | XP_014986844.1 | 979 | 103 | |||
XM_015131360.2 | XP_014986846.1 | 628 | 103 | |||
GenBank mRNA | JU335913.1 | - | 722 | 103 | ||
JU476790.1 | - | 681 | 103 | |||
JV658753.1 | - | 772 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |