Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265727.1 | NP_001252656.1 | 1384 | 104 | ||
XM_015146907.2 | XP_015002393.2 | 1670 | 104 | |||
XM_015146912.2 | XP_015002398.1 | 1681 | 104 | |||
XM_028836339.1 | XP_028692172.1 | 1814 | 104 | |||
GenBank mRNA | JU350388.1 | - | 1124 | 104 | ||
JV636444.1 | - | 1384 | 104 | |||
JV726621.1 | - | 1425 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |