Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266435.1 | NP_001253364.1 | 1440 | 106 | ||
GenBank mRNA | JU332444.1 | - | 1440 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |