Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194502.3 | NP_001181431.2 | 338 | 66 | ||
GenBank mRNA | JU327025.1 | - | 315 | 66 | ||
JV049568.1 | - | 319 | 66 | |||
JV565029.1 | - | 326 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |