Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194445.2 | NP_001181374.1 | 417 | 68 | ||
GenBank mRNA | JU472957.1 | - | 316 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |