Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193749.1 | NP_001180678.1 | 581 | 96 | ||
GenBank mRNA | JU334140.1 | - | 418 | 96 | ||
JV046837.1 | - | 434 | 96 | |||
JV579453.1 | - | 450 | 96 | |||
JV649239.1 | - | 460 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |