Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266242.1 | NP_001253171.1 | 396 | 133 | ||
XM_015114484.2 | XP_014969970.1 | 274 | 133 | |||
GenBank mRNA | JU326041.1 | - | 328 | 133 | ||
JV045743.1 | - | 396 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |