Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193806.1 | NP_001180735.1 | 158 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |