Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM255A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193758.1 | NP_001180687.1 | 454 | 136 | ||
XM_001085757.2 | XP_001085757.1 | 454 | 136 | |||
XM_028841669.1 | XP_028697502.1 | 454 | 136 | |||
GenBank mRNA | JU335206.1 | - | 346 | 136 | ||
JU335207.1 | - | 346 | 136 | |||
JV047488.1 | - | 272 | 136 | |||
JV636245.1 | - | 421 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |