Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02874920_m1
See other PPP6R3 GE Assays ›
Gene Symbol:
PPP6R3
Gene Name:
protein phosphatase 6 regulatory subunit 3
Gene Aliases:
EGK_05908, SAPS3
Chromosome Location:
Chr.14: 6149374 - 6307267 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02874920_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PPP6R3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015113490.2
XP_014968976.1
2014
124
XM_015113492.2
XP_014968978.1
1996
124
XM_015113495.2
XP_014968981.1
1927
124
XM_028833114.1
XP_028688947.1
1721
124
XM_028833115.1
XP_028688948.1
1688
124
XM_028833116.1
XP_028688949.1
1739
124
XM_028833117.1
XP_028688950.1
1721
124
XM_028833118.1
XP_028688951.1
1652
124
XM_028833119.1
XP_028688952.1
1634
124
XM_028833120.1
XP_028688953.1
1688
124
XM_028833121.1
XP_028688954.1
1688
124
XM_028833122.1
XP_028688955.1
1601
124
XM_028833123.1
XP_028688956.1
1586
124
XM_028833124.1
XP_028688957.1
1652
124
XM_028833125.1
XP_028688958.1
1739
124
XM_028833126.1
XP_028688959.1
1634
124
XM_028833127.1
XP_028688960.1
1634
124
XM_028833128.1
XP_028688961.1
2014
124
XM_028833129.1
XP_028688962.1
2363
124
XM_028833130.1
XP_028688963.1
2112
124
XM_028833131.1
XP_028688964.1
2079
124
XM_028833132.1
XP_028688965.1
2111
124
XM_028833133.1
XP_028688966.1
1940
124
XM_028833134.1
XP_028688967.1
1601
124
XM_028833136.1
XP_028688969.1
1535
124
XM_028833137.1
XP_028688970.1
1499
124
GenBank mRNA
JU321666.1
-
1799
124
JU321700.1
-
1886
124
JU321705.1
-
1886
124
JU321706.1
-
1886
124
JU321718.1
-
1886
124
JU471603.1
-
1996
124
JU471604.1
-
1909
124
JV047549.1
-
1947
124
JV047550.1
-
1860
124
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Macaca mulatta
Panther Classification:
Molecular Function -
phosphatase modulator
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-r642z:80/10.239.167.51:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0