Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085366.4 | XP_001085366.2 | 1452 | 149 | ||
XM_015136335.2 | XP_014991821.2 | 1514 | 149 | |||
XM_028847582.1 | XP_028703415.1 | 1474 | 149 | |||
XM_028847583.1 | XP_028703416.1 | 1454 | 149 | |||
GenBank mRNA | JU484013.1 | - | 1572 | 149 | ||
JV711483.1 | - | 323 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |