Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257527.1 | NP_001244456.1 | 287 | 120 | ||
XM_015133877.1 | XP_014989363.1 | 439 | 120 | |||
XM_028844996.1 | XP_028700829.1 | 439 | 120 | |||
GenBank mRNA | JU335344.1 | - | 287 | 120 | ||
JU476296.1 | - | 271 | 120 | |||
JU476297.1 | - | 271 | 120 | |||
JU476298.1 | - | 271 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |