Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115228.2 | XP_014970714.1 | 3463 | 132 | ||
XM_015115229.2 | XP_014970715.1 | 3460 | 132 | |||
XM_015115230.2 | XP_014970716.1 | 3436 | 132 | |||
XM_015115231.2 | XP_014970717.1 | 3421 | 132 | |||
XM_015115232.2 | XP_014970718.1 | 3433 | 132 | |||
XM_015115233.2 | XP_014970719.1 | 3310 | 132 | |||
XM_015115234.2 | XP_014970720.1 | 3298 | 132 | |||
XM_015115236.2 | XP_014970722.1 | 3283 | 132 | |||
XM_015115237.2 | XP_014970723.1 | 3268 | 132 | |||
XM_015115239.2 | XP_014970725.1 | 3394 | 132 | |||
GenBank mRNA | JU335540.1 | - | 3414 | 132 | ||
JU476467.1 | - | 3414 | 132 | |||
JV719156.1 | - | 3419 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |